2-Iodomesitylene ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 2,4,6-Trimethyliodobenzene, 1-Iodo-2,4,6-trimethylbenzene, 2-Iodo-1,3,5-trimethylbenzene, Iodomesitylene

I0478-25G I0478-5G
TCIAI0478-25GEA 1350 NOK
TCIAI0478-25G TCIAI0478-5G
2-Iodomesitylene ≥98.0% (by GC)
2-Iodomesitylene
Formel: C₉H₁₁I
MW: 246,09 g/mol
Kokepunkt: 250°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00013707
CAS-nummer: 4028-63-1
EINECS: 223-709-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dd0c518f5b6f40a491579f46c0f2db096bceb5d&vid=1dd0c518f5b6f40a491579f46c0f2db096bceb5d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Melting point 30.0 to 33.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR