5-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)

Leverandør: TCI
Warning

Synonymer: 5-Fluoro-2-methoxybenzeneboronic acid

F0775-25G F0775-5G
TCIAF0775-25GEA 2500 NOK
TCIAF0775-25G TCIAF0775-5G
5-Fluoro-2-methoxyphenylboronic acid (contains varying amounts of Anhydride)
5-Fluoro-2-methoxyphenylboronic acid
Formel: C₇H₈BFO₃
MW: 169,95 g/mol
Smeltepunkt: 158°C
MDL nummer: MFCD01863526
CAS-nummer: 179897-94-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e36c361d2d745aae54254cdea4616ee020e5de3&vid=8e36c361d2d745aae54254cdea4616ee020e5de3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR