L(-)-Methioninol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: (S)-2-Amino-4-(methylthio)butan-1-ol, L-Methioninol

M0929-1G M0929-100MG
TCIAM0929-1GEA 1750 NOK
TCIAM0929-1G TCIAM0929-100MG
L(-)-Methioninol ≥98.0% (by GC)
L(-)-Methioninol
Formel: C₅H₁₃NOS
MW: 135,2 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00004735
CAS-nummer: 2899-37-8
EINECS: 220-788-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d9cc9a2dd1b20ba0ca58dd2de0e2d0f4cffb93b7&vid=d9cc9a2dd1b20ba0ca58dd2de0e2d0f4cffb93b7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 32.0 to 36.0 deg-C
Specific rotation [a]20/D -13.0 to -15.0 deg (C=1.4, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR