2,4-Dimethyl-3-ethylpyrrole ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Cryptopyrrole, 3-Ethyl-2,4-dimethylpyrrole, Kryptopyrrole

E0555-1G E0555-5G
TCIAE0555-1GEA 485 NOK
TCIAE0555-1G TCIAE0555-5G
2,4-Dimethyl-3-ethylpyrrole ≥97.0% (by GC)
2,4-Dimethyl-3-ethylpyrrole
Formel: C₈H₁₃N
MW: 123,2 g/mol
Flash punkt: 71°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00005222
CAS-nummer: 517-22-6
EINECS: 208-234-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfb31d7cd41009734724ac4ccd717e35ac38c537&vid=cfb31d7cd41009734724ac4ccd717e35ac38c537&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR