2-Phenylisobutyric acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Methyl-2-phenylpropanoic acid, α,α-Dimethylphenylacetic acid

P1409-25G P1409-250G
TCIAP1409-25GEA 500 NOK
TCIAP1409-25G TCIAP1409-250G
2-Phenylisobutyric acid ≥98.0% (by GC, titration analysis)
2-Phenylisobutyric acid
Formel: C₁₀H₁₂O₂
MW: 164,21 g/mol
MDL nummer: MFCD00014332
CAS-nummer: 826-55-1
EINECS: 212-556-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26de338dd5a08ff11061fd841248b6e8ae7644b7&vid=26de338dd5a08ff11061fd841248b6e8ae7644b7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 77.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR