Scopoletin (7-hydroxy-6-methoxycumarin) ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one

S0367-1G S0367-100MG
TCIAS0367-1GEA 4775 NOK
TCIAS0367-1G TCIAS0367-100MG
Scopoletin (7-hydroxy-6-methoxycumarin) ≥98.0% (by HPLC, titration analysis)
Scopoletin (7-hydroxy-6-methoxycumarin)
Formel: C₁₀H₈O₄
MW: 192,1681 g/mol
MDL nummer: MFCD00006872
CAS-nummer: 92-61-5
EINECS: 202-171-9
Merck Indeks: 14,08408
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95fa88669f6f4b5e23f3bea5cd689cab57b1a937&vid=95fa88669f6f4b5e23f3bea5cd689cab57b1a937&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 203.0 to 210.0 deg-C
Solubility in hot EtOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR