Triphenylbismuth carbonate ≥98.0% (by titrimetric analysis)

Leverandør: TCI

T1835-1G T1835-5G
TCIAT1835-1GEA 1375 NOK
TCIAT1835-1G TCIAT1835-5G
Triphenylbismuth carbonate ≥98.0% (by titrimetric analysis)
Triphenylbismuth carbonate
Formel: C₁₉H₁₅BiO₃
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00209622
CAS-nummer: 47252-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d32253eff06bb0ffb138cea4c2ebe9eafe269e5&vid=1d32253eff06bb0ffb138cea4c2ebe9eafe269e5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Water max. 4.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR