Jasmonic acid (mixture of isomers) ≥85.0% (by GC)
Leverandør: TCI
Synonymer:
3-(Carboxymethyl)-2-(2-pentenyl)cyclopentanone, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid
J0004-1G
J0004-5G
TCIAJ0004-1GEA
1750
NOK
TCIAJ0004-1G
TCIAJ0004-5G
Jasmonic acid (mixture of isomers) ≥85.0% (by GC)
Jasmonic acid (mixture of isomers)
Formel:
C₁₂H₁₈O₃ MW: 210,27 g/mol |
CAS-nummer:
221682-41-3 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 85.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...