2-Aminoisonicotinic acid ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Amino-4-pyridinecarboxylic acid, 2-Aminopyridine-4-carboxylic acid

A2332-1G A2332-5G
TCIAA2332-1GEA 1025 NOK
TCIAA2332-1G TCIAA2332-5G
2-Aminoisonicotinic acid ≥97.0% (by HPLC, titration analysis)
2-Aminoisonicotinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Smeltepunkt: 290°C
MDL nummer: MFCD00233714
CAS-nummer: 13362-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=075e055c30eea7e22d893e7230cad8e922fe13b0&vid=075e055c30eea7e22d893e7230cad8e922fe13b0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystalline
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR