Triphenylbismuth difluoride ≥97.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
T2657-1G
T2657-5G
TCIAT2657-1GEA
1325
NOK
TCIAT2657-1G
TCIAT2657-5G
Triphenylbismuth difluoride ≥97.0% (by titrimetric analysis)
Triphenylbismuth difluoride
Formel:
C₁₈H₁₅BiF₂ Smeltepunkt: 159°C Lagringstemperatur: Kjøleskap |
CAS-nummer:
2023-48-5 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...