5-Chloro-2-(2-chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Synonymer:
2-Amino-2',4-dichlorodiphenyl ether, 2-Amino-2,4-dichlorodiphenyl ether, 5-Chloro-2-(2-chlorophenoxy)aniline
A2224-25G
A2224-5G
TCIAA2224-25GEA
4100
NOK
TCIAA2224-25G
TCIAA2224-5G
5-Chloro-2-(2-chlorophenoxy)aniline ≥98.0% (by GC, titration analysis)
5-Chloro-2-(2-chlorophenoxy)aniline
Formel:
C₁₂H₉Cl₂NO MW: 254,1120 g/mol Kokepunkt: 210°C |
MDL nummer:
MFCD00025218 CAS-nummer: 56966-48-4 EINECS: 260-479-9 |
Spesifikasjon Testresultater
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...