Palladium 5% on activated carbon paste (50% water) A503023-5

Supplier: Thermo Fisher Scientific

11694.06 11694.14 11694.22
11694.06EA 1300 NOK
11694.06 11694.14 11694.22
Palladium 5% on activated carbon paste (50% water) A503023-5
Palladium

CAS No.: 7440-05-3 Appearance: Black Odour: Odourless MDL No.: MFCD03457879


Applications: Debenzylation

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR