Palladium 5% on calcium carbonate, powder for use as a catalyst, reduced

Supplier: Thermo Fisher Scientific

11723.06 11723.14 11723.22
11723.06EA 1300 NOK
11723.06 11723.14 11723.22
Palladium 5% on calcium carbonate, powder for use as a catalyst, reduced
Palladium

CAS No.: 7440-05-3 Odour: Odourless MDL No.: MFCD03427452

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR