Palladium 5% on strontium carbonate, powder, reduced

Supplier: Thermo Fisher Scientific

39819.04 39819.09 39819.18
39819.04EA 810 NOK
39819.04 39819.09 39819.18
Palladium 5% on strontium carbonate, powder, reduced
Palladium
Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00192595
CAS Number: 7440-05-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR