2-Bromethylamine hydrobromide ≥98.0%, white crystalline powder

Supplier: MP Biomedicals
Warning

0215487525 0215487580
ICNA0215487525EA 300 NOK
ICNA0215487525 ICNA0215487580
2-Bromethylamine hydrobromide ≥98.0%, white crystalline powder
2-Bromethylamine hydrobromide

2-Bromoethylamine is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone.


Caution: Research Use Only (RUO).

Formula: BrCH₂CH₂NH₃Br
Melting Pt: 342…345° F (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR