Drum spill kits
Supplier: Brady
813869 813870 813871
BRDY813869EA 6500 NOK
BRDY813869 BRDY813870 BRDY813871
Drum spill kits
Spill Control Materials
The drum spill kit is designed for medium sized spills.
  • Water and chemical resistant drum protects sorbents
  • Protects sorbents against moisture, dirt and damage
  • Drum can also be used as shipping container for used sorbents
  • Easy access to products with screw top lid
Delivery information: Supplied with 12 pads (410×510 mm), 3 socs (Ø 7,6×3660 mm), 2 pillows (430×480 mm), 1 pair of gloves, 1 pair of goggles and 3 disposal bags.
Order Now
SPESIFIKASJONER
Environmentally Preferable
Learn more
About VWR
Les mer About VWR