Palladium 5% on barium sulphate, powder hydrogenation catalyst (oxidised form), unreduced

Supplier: Thermo Fisher Scientific

21162.09 21162.18
21162.09EA 2575 NOK
21162.09 21162.18
Palladium 5% on barium sulphate, powder hydrogenation catalyst (oxidised form), unreduced
Palladium

Appearance: Pale brown MDL No.: MFCD03613605


Hydrodehalogenation, Rosenmund reduction, asymetric hydrogenation

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR