Magnesium chloride hexahydrate USP, Multi-Compendial, J.T.Baker®

Supplier: Avantor


Management of Change (MOC) category = HR

2449-01EA 2775 NOK
2449-01 2449-07
Magnesium chloride hexahydrate USP, Multi-Compendial, J.T.Baker®
Magnesium chloride hexahydrate
Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Meets B.P. Chemical Specifications
GMP Manufactured Product
Meets E.P. Chemical Specifications
Meets U.S.P Requirements
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical Passes Test
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (1 in 20) 4.5 - 7.0
USP - Insoluble Matter ≤ 0.005 %
USP - Sulfate (SO₄) ≤ 0.005 %
USP - Barium (Ba) Passes Test
USP - Calcium (Ca) ≤ 0.01 %
USP - Potassium (K) Passes Test
USP - Assay 98.0 - 101.0 %
USP - Endotoxin Concentration (EU/g) ≤ 2.5
EP/BP - Assay 98.0 - 101.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Acidity or Alkalinity Passes Test
EP/BP - Bromide (Br) ≤ 500 ppm
EP/BP - Sulfate (SO₄) ≤ 100 ppm
EP/BP - Calcium (Ca) ≤ 0.1 %
EP/BP - Iron (Fe) ≤ 10 ppm
EP/BP - Water (H₂O) 51.0 - 55.0 %
No Class 1,2,3 or other solvents are used or produced in the 51.0 - 55.0 %
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR