4'-Ethoxyacetophenone ≥98%

Supplier: Thermo Fisher Scientific

A11031.06 A11031.14 A11031.22
A11031.06EA 460 NOK
A11031.06 A11031.14 A11031.22
4'-Ethoxyacetophenone ≥98%
4'-Ethoxyacetophenone

Purity: 98% CAS No.: 1676-63-7 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 134°(273°F) Melting Point: 35-38° Boiling Point: 145-146°/13mm Beilstein Reference: 636783 MDL No.: MFCD00009095

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 145…146 °C (13 mmHg)
Melting Pt: 35…38 °C
Flash Pt: 134 °C (273 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009095
CAS Number: 1676-63-7
EINECS: 216-825-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR