2-Bromethylamine hydrobromide ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

85987.260EA 2200 NOK
85987.260 85987.360
2-Bromethylamine hydrobromide ≥99%, GPR RECTAPUR®
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2

Order Now

Specification Test Results

Assay Min. 99 %
Melting point 171 --> 175 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR