Palladium 5% on carbon, powder Type 490

Supplier: Thermo Fisher Scientific
Warning

H33217.09 H33217.18
H33217.09EA 13075 NOK
H33217.09 H33217.18
Palladium 5% on carbon, powder Type 490
Palladium
Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR