Platinum nominally 20% on carbon black

Supplier: Thermo Fisher Scientific

Synonyms: Platinium

47381.06 47381.03
47381.06EA 8325 NOK
47381.06 47381.03
Platinum nominally 20% on carbon black
Platinum
Formula: Pt
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR