2-Bromethylamine hydrobromide 98%

Supplier: Apollo Scientific
Warning

OR1160-1KG OR1160-500G OR1160-100G
APOSOR1160-1KGEA 1850 NOK
APOSOR1160-1KG APOSOR1160-500G APOSOR1160-100G
2-Bromethylamine hydrobromide 98%
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR