2-Bromethylamine hydrobromide 99%

Supplier: Thermo Fisher Scientific
Warning

106930250 106931000 106930010 106930025 106932500 106935000
ACRO106930250EA 405 NOK
ACRO106930250 ACRO106931000 ACRO106930010 ACRO106930025 ACRO106932500 ACRO106935000
2-Bromethylamine hydrobromide 99%
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration Argentometric ≥98.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR