2-Bromethylamine hydrobromide ≥98%

Supplier: Thermo Fisher Scientific
Warning

A17625.18 A17625.0B A17625.30
A17625.18EA 415 NOK
A17625.18 A17625.0B A17625.30
2-Bromethylamine hydrobromide ≥98%
2-Bromethylamine hydrobromide

Purity: 98+% CAS No.: 2576-47-8 Molecular Formula: BrCH2CH2NH2.HBr Formula Weight: 204.90 Melting Point: 172-176° Beilstein Reference: 3607605

Formula: BrCH₂CH₂NH₃Br
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR