Hydrochloric acid 30%, Suprapur® for trace analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

1.00318.0250 1.00318.1000 1.00318.0500 1.00318.2500
1.00318.0250EA 580 NOK
1.00318.0250 1.00318.1000 1.00318.0500 1.00318.2500
Hydrochloric acid 30%, Suprapur® for trace analysis, Supelco®
Hydrochloric acid
Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay (acidimetric) ≥ 30.0 %
Colour ≤ 10 Hazen
Bromide (Br) ≤ 10 ppm
Free chlorine (Cl) ≤ 300 ppb
Phosphate (PO₄) ≤ 10 ppb
Sulphate (SO₄) ≤ 300 ppb
Sulfite (SO₃) ≤ 500 ppb
Ag (Silver) ≤ 1.0 ppb
Al (Aluminium) ≤ 1.0 ppb
As (Arsenic) ≤ 1.0 ppb
Au (Gold) ≤ 1.0 ppb
Ba (Barium) ≤ 1.0 ppb
Be (Beryllium) ≤ 1.0 ppb
Bi (Bismuth) ≤ 1.0 ppb
Ca (Calcium) ≤ 5.0 ppb
Cd (Cadmium) ≤ 0.5 ppb
Co (Cobalt) ≤ 1.0 ppb
Cr (Chromium) ≤ 1.0 ppb
Cu (Copper) ≤ 1.0 ppb
Fe (Iron) ≤ 5.0 ppb
Ga (Gallium) ≤ 0.5 ppb
Ge (Germanium) ≤ 10.0 ppb
Hg (Mercury) ≤ 2.0 ppb
In (Indium) ≤ 0.5 ppb
K (Potassium) ≤ 10.0 ppb
Li (Lithium) ≤ 0.5 ppb
Mg (Magnesium) ≤ 2.0 ppb
Mn (Manganese) ≤ 0.5 ppb
Mo (Molybdenum) ≤ 0.5 ppb
NH₄ (Ammonium) ≤ 500 ppb
Na (Sodium) ≤ 10.0 ppb
Ni (Nickel) ≤ 1.0 ppb
Pb (Lead) ≤ 0.5 ppb
Pt (Platinum) ≤ 0.5 ppb
Sb (Antimony) ≤ 0.5 ppb
Sn (Tin) ≤ 1.0 ppb
Sr (Strontium) ≤ 0.5 ppb
Ti (Titanium) ≤ 1.0 ppb
Tl (Thallium) ≤ 0.5 ppb
V (Vanadium) ≤ 1.0 ppb
Zn (Zinc) ≤ 1.0 ppb
Zr (Zirconium) ≤ 0.5 ppb
Residue on ignition (as sulphate) ≤ 2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR