Hydrogen peroxide 30%, Suprapur® for trace analysis, Supelco®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

1.07298.0250 1.07298.1000 1.07298.0500
1.07298.0250EA 1930 NOK
1.07298.0250 1.07298.1000 1.07298.0500
Hydrogen peroxide 30%, Suprapur® for trace analysis, Supelco®
Hydrogen peroxide
Formula: H₂O₂
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -26 °C
Boiling Point 107 °C (1013 hPa)
Vapor Pressure 18 hPa (20 °C)
Density 1.11 g/cm3 (20 °C)
pH 2 - 4 (H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR