Potassium chloride 99.0-100.5% Ph. Eur., USP

Supplier: VWR Chemicals

26760.295EA 735 NOK
26760.295 26760.364 26760.466
Potassium chloride 99.0-100.5% Ph. Eur., USP
Potassium chloride

Appearance: White or almost white, crystalline powder or colourless crystals.


Caution: Not for use as Active Pharmaceutical Ingredient (API)

Formula: KCl
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) ≤ 10 ppm
Residual solvents Unlikely by manuf.process
Residues of metal catalysts or reagents Unlikely by manuf.process
As (Arsenic) ≤ 2 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Ni (Nickel) ≤ 1 ppm
Pb (Lead) ≤ 2 ppm
V (Vanadium) ≤ 6 ppm
Conforms to Ph.Eur./USP Passes test
Ph.Eur. specifications: .
Assay (dried substance) [Ph.Eur.] 99.0 - 100.5 %
Identification A Passes test USP
Identification B Passes test USP
Appearance of solution Passes test Ph.Eur.
Acidity or alkalinity Passes test USP
Br (Bromide) [Ph.Eur.] ≤ 0.1 %
I (Iodide) Passes test Ph.Eur.
SO4 (Sulphate) [Ph.Eur.] ≤ 300 ppm
Fe (Iron) [Ph.Eur.] ≤ 20 ppm
Mg and alk. earth metals(as Ca)[Ph.Eur.] ≤ 200 ppm
Loss on drying (105°C; 3 h) [Ph.Eur.] ≤ 1.0 %
USP specifications: .
Assay (calculated on dried basis) [USP] 99.0 - 100.5 %
Na (Sodium) Passes test USP
I (Iodide) [USP] ≤ 0.005 %
Br (Bromide) [USP] ≤ 0.1 %
Calcium and magnesium Passes test USP
Loss on drying (105°C; 2 h) [USP] ≤ 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR