Sodium chloride 99.0-100.5% Ph. Eur., USP, BP

Supplier: VWR Chemicals

27808.297EA 540 NOK
27808.297 27808.366 27808.468
Sodium chloride 99.0-100.5% Ph. Eur., USP, BP
Sodium chloride

Appearance: White or almost white, crystalline powder or colourless crystals or white or almost white pearls.

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) Max. 5 ppm
Residual solvents Passes test
Cd (Cadmium) Max. 1 ppm
Co (Cobalt) Max. 1 ppm
Hg (Mercury) Max. 2 ppm
Ni (Nickel) Max. 1 ppm
Pb (Lead) Max. 2 ppm
V (Vanadium) Max. 1 ppm
Conforms to Ph.Eur./USP/BP Passes test
Ph.Eur./BP specifications: Passes test
Assay (dried substance) [Ph.Eur./BP] 99.0 --> 100.5 %
Identification A Passes test USP
Identification B Passes test USP
Appearance of solution Passes test USP
Acidity or alkalinity Passes test USP
Br (Bromide) [Ph.Eur./BP] Max. 100 ppm
Ferrocyanides Passes test USP
I (Iodide) Passes test USP
NO2 (Nitrite) Passes test Ph.Eur./BP
PO4 (Phosphate) [Ph.Eur./BP] Max. 25 ppm
SO4 (Sulphate) [Ph.Eur./BP] Max. 200 ppm
As (Arsenic) [Ph.Eur./BP] Max. 1 ppm
Ba (Barium) Passes test USP
Fe (Iron) [Ph.Eur./BP] Max. 2 ppm
Mg + alk. earth metals (as Ca) [Ph.Eur./BP] Max. 100 ppm
Loss on drying (105°C; 2 h) [Ph.Eur./BP] Max. 0.5 %
USP specifications:
Assay (calculated on dried basis) [USP] 99.0 --> 100.5 %
As (Arsenic) [USP] Max. 1 ppm
Fe (Iron) [USP] Max. 2 ppm
Limit of bromides [USP] Max. 100 ppm
Limit of phosphates [USP] Max. 25 ppm
Mg and alkaline earth metals(as Ca)[USP] Max. 100 ppm
NO2 (Nitrite) (A 354 nm) [USP] Max. 0.01
SO4 (Sulphate) [USP] Max. 200 ppm
Loss on drying (105°C; 2 h) [USP] Max. 0.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR