VWR® SPILL-X® Spill Kits, Mobile
Supplier: VWR Chemicals
121-0006EA 28725 NOK
121-0006 121-0007
VWR® SPILL-X® Spill Kits, Mobile
Spill Control Materials
Developed to economically treat small spills, such as those that can occur in a laboratory environment. Provides easy access to the appropriate control agent for a particular type of chemical spill.
  • Treats hazardous spills rather than just repackaging them
  • Safe, easy to use, with the highest per volume absorption capacity
  • Neutralises and solidifies acids and caustics for easy waste disposal (SPILL-X-A®, SPILL-X-C®)
  • Absorbs solvents and raises the flash point above 60 °C, without producing toxic by-products (SPILL-X-S®)
  • SPILL-X® case comes with portable,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Learn more
About VWR
Les mer About VWR