2-Bromethylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI
Warning

B0591-25G B0591-500G B0591-100G
TCIAB0591-25GEA 285 NOK
TCIAB0591-25G TCIAB0591-500G TCIAB0591-100G
2-Bromethylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
2-Bromethylamine hydrobromide
Formula: BrCH₂CH₂NH₃Br
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=649f03a9cf7c4bff37af44e912d6021a536574de&vid=649f03a9cf7c4bff37af44e912d6021a536574de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 173.0 to 177.0 deg-C
Solubility in Water almost transparency

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR