Non-aqueous DPX mounting medium
Leverandør: Merck
1.00579.0500
1.00579.0500EA 752 NOK
1.00579.0500
Non-aqueous DPX mounting medium
Farger Microscopy and staining reagents
DPX new is a synthetic non aqueous mounting medium for microscopy. In its formulation, the teratogenic plasticiizer dibutyl phthalate (DBP) has been avoided, which renders the composition safer for the environment and more user friendly than the conventionally used mounting agents such as DPX.
Preparative capacity (500 ml): 2500 slides