Mounting medium, kaiser's glycerol gelatine, phenol-free

Leverandør: Merck

1.08635.0100
1.08635.0100EA 625 NOK
1.08635.0100
Mounting medium, kaiser's glycerol gelatine, phenol-free
Farger Microscopy and staining reagents

Kaiser's glycerol gelatin is an aqueous mounting medium for microscopy and works well with specimens prepared for enzyme, lipid, and immunohistochemistry determinations.


  • Density: 1,117 to 1,120 g/cm³ (40 °C)
  • Storage: +15 °C to +25 °C


The preservative phenol is replaced with a proprietary compound that is more environmentally friendly. This affords the unique advantage of eliminating phenol fumes when the medium is heated before use. It is developed specifically for use in histological applications is essential in the support of formal quality assurance programs to monitor staining performance in pathology laboratories.


Preparation Capacity: 400 slides.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR