Horizontal laminar flow cabinets, Aeolus Series, Telstar

Leverandør: Azbil Telstar Technologies
1511967 1511968 1511969 1511970
135-2608EA 0 NOK
135-2608 135-2609 135-2610 135-2611
Horizontal laminar flow cabinets, Aeolus Series, Telstar
Avtrekksskap og kabinetter Laminert flowbenk
Aeolus H Serie is a high quality horizontal laminar flow workbench range for handling non-hazardous materials that require a particle free environment. These cabinets offer the highest product protection. Aeolus H horizontal laminar flow benches, have been specially designed to work in hospitals, pharmacies, IVF centres, food control, horticulture in vitro cultures, and also in electronics, optics, micromechanics, plastic Industries, etc. Horizontal laminar flow systems allow operation in sterile and particle free conditions thanks to the continuous flushing of the working area by means of aMore Product Information
Bestill nå

SPESIFIKASJONER

Modell Aeolus H 3 Aeolus H 4 Aeolus H 5 Aeolus H 6
Environmentally Preferable
Vifter EC High efficiency Centrifugal EC High efficiency Centrifugal EC High efficiency Centrifugal EC High efficiency Centrifugal
Lufthastighet 0,45 m/s 0,45 m/s 0,45 m/s 0,45 m/s
Lys ≥800 lux ≥800 lux ≥900 lux ≥900 lux
Støynivå 57 dB(A) 58 dB(A) 59 dB(A) 60 dB(A)
Filtre HEPA: Efficiency 99, 995%, EN 1822 by MPPS method (99,999% D.O.P. test @ 0,3 μm particles)
Pre-filter efficient (EN 779): G3
HEPA: Efficiency 99, 995%, EN 1822 by MPPS method (99,999% D.O.P. test @ 0,3 μm particles)
Pre-filter efficient (EN 779): G3
HEPA: Efficiency 99, 995%, EN 1822 by MPPS method (99,999% D.O.P. test @ 0,3 μm particles)
Pre-filter efficient (EN 779): G3
HEPA: Efficiency 99, 995%, EN 1822 by MPPS method (99,999% D.O.P. test @ 0,3 μm particles)
Pre-filter efficient (EN 779): G3
Utvendig B×D×H 944×872×1212 mm 1249×872×1212 mm 1554×872×1212 mm 1859×872×1212 mm
Innvendig B×D×H 925×598×780 mm 1230×598×780 mm 1535×598×780 mm 1840×598×780 mm
Vekt 106 kg 134 kg 150 kg 171 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR