VWR®, D.F.B (Decalcifying solution - H.K. Kristenson) Histological grade
Leverandør: VWR Chemicals
352432CEA 1575 NOK
352432C
VWR®, D.F.B (Decalcifying solution - H.K. Kristenson) Histological grade
Farger Microscopy and staining reagents
Decalcifier according to Kristenson.
  • pH (25°C) 2.20 --> 2.70