PCR plates, 384-well
Leverandør: Brand
781345
391-5711EA 7725 NOK
391-5711
PCR plates, 384-well
Mikroplater PCR-plater
PP. The 40 μl wells can hold sample volumes of 2 to 30 μl. This helps to reduce reagent costs and shortens cycle times. The plates can be filled using multichannel pipettes or robotic systems.
  • Ultra-thin walled design to facilitate constant, rapid and precise heat transfer
  • Smooth vessel interior minimises the binding of enzymes and nucleic acid to the walls
  • Embossed alphanumeric coding
  • Raised well rims protect against cross-contamination and provide a tight seal in combination with matching sealing mat
  • Free from DNase, RNase and DNA
  • Autoclavable at 121 °C (