Epoxy embedding medium, hardener MNA, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
45347-1L-F
45347-1L-FEA 1370 NOK
45347-1L-F
Epoxy embedding medium, hardener MNA, Sigma-Aldrich®
Farger Microscopy and staining reagents
Epoxy embedding medium is generally used for biological tissues during study by electron microscopy. Usually, it consists of three components; Epon 812 resin, an anhydride hardener and an accelerator
  • Epon™ MNA substitute
Application
It was used during transmission electron microscopy done for the analysis of epithelial cells; during a study performed to understand the epithelial cell growth and stratification in minimal culture conditions.
Epon is a trademark of Hexion, Inc.