NBB®-B-AM, Broth
Leverandør: DOEHLER GROUP
204706782
710-0738EA 2850 NOK
710-0738
NBB®-B-AM, Broth
Mikrobiologiske medier Media, preparerte for mikrobiologi
Culture medium for qualitative detection of indicator microorganisms using swab sampling with sterile swabs as part of microbiological facility monitoring. Under its NBB® brand, Döhler offers innovative culture media for the fast and reliable detection of beverage spoiling microorganisms, such as Lactobacilli, Pectinatus and Megasphaera, in all samples that arise in breweries, as well as for the wine industry. NBB®-B-AM is a liquid culture medium for detection of biofilm indicator microorganisms.
  • Simple hygiene monitoring for weak point analyses and trending
  • Sampling by means of smear swabs at critical points throughout the production process
  • Direct use of liquid medium
  • Simple, visual screening by an indicator change from red to yellow