PCR plates for qPCR, 24-well and 48-well
Leverandør: Brand
781411 781415 781412 781416
731-0014EA 1550 NOK
731-0014 731-0015 731-0165 731-0166
PCR plates for qPCR, 24-well and 48-well
Mikroplater PCR-plater
PP, non skirted. Manufactured under cleanroom conditions, and made from pure, high quality materials (without additives) these PCR plates are suitable for use with most commercially available thermal cyclers.
  • Ultra-thin walled design to facilitate constant, rapid and precise heat transfer
  • Smooth vessel interior minimises the binding of enzymes and nucleic acid to the walls
  • Alphanumeric coding embossed and stamped in blue
  • Raised well rims protect against cross-contamination and provide a tight seal in combination with matching sealing mat
  • Free from DNase,