PCR plates, 96-well, semi-skirted, flat deck, barcoded
Leverandør: Thermo Fisher Scientific
BC-1400 BC-1400/W BC-2400 BC-2400/W
ADVABC-1400EA 1550 NOK
ADVABC-1400 ADVABC-1400/W 732-1494 732-1495
PCR plates, 96-well, semi-skirted, flat deck, barcoded
Mikroplater PCR-plater
These PP plates are directly compatible with all standard platforms including sequencers with no adapters necessary.
  • Directly compatible with all major major thermal cyclers, including ABI platforms including sequencers with no adapters necessary
  • Flat plate deck facilitates sealing and handling
  • Cut corner: A12
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • Black alpha-numeric lettering allows for easy well identification
  • White plates are