VWR® Rigid qPCR Plates, 96-Well, with White Wells
Leverandør: VWR Collection
732-4403EA 815 NOK
732-4403 732-4405
VWR® Rigid qPCR Plates, 96-Well, with White Wells
Mikroplater PCR-plater
Polycarbonate frame is rigid and flat to facilitate a firm grip by robotic arms and ensure reliable placement. Polypropylene wells provide optimal thermal transfer and higher reaction efficiency.
  • White well options enhance the fluorescence signal during qPCR
  • Wells are slightly raised to ensure evaporation-proof sealing with film, foil, or strip caps
  • ANSI/SBS footprint and well spacing
Vibrant colours offer a visual distinction between different samples or experiments.