96-Well clear polypropylene PCR microplate, Corning® Thermowell® GOLD
Leverandør: Corning
3758
734-4137EA 7075 NOK
734-4137
96-Well clear polypropylene PCR microplate, Corning® Thermowell® GOLD
Mikroplater PCR-plater
These half skirted 96 well PCR microplates feature a rigid construction that is ideal for automation.
  • Clear medical grade PP is suitable for RT-PCR
  • Uniform wall thinness assures efficient heat transfer while minimizing well-to-well variability
  • 100% tested for well integrity and DNase- and RNase-free
  • Nonsterile, without lids
  • With two generic bar code labels per plate