RenLam® M-1 for microscopy, suitable as a embedding medium, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
78711-1L 78711-250ML
78711-1LEA 1320 NOK
78711-1L 78711-250ML
RenLam® M-1 for microscopy, suitable as a embedding medium, Sigma-Aldrich®
Farger Microscopy and staining reagents
To be used with SIAL 69571 Ren HY 956 (Hardener for Epoxy Embedding Medium)
NB: Renlam is a registered trademark of Huntsman Advanced Materials Inc.