VWR® Gurr™ Histological Fixatives, AFA
Leverandør: VWR Chemicals
81009.362EA 3650 NOK
81009.362
VWR® Gurr™ Histological Fixatives, AFA
Farger Microscopy and staining reagents
Acidified alcoholic formalin for the fixation of glycogen.
Contains acetic acid, ethanol and formaldehyde.