VWR® Fixing Solution
Leverandør: VWR Chemicals
87970.260EA 2175 NOK
87970.260
VWR® Fixing Solution
Farger Microscopy and staining reagents
Fixing solution with 0,02% formaldehyde in methanol 50% (v/v)).
Bestillingsinformasjon: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.