ROX Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Leverandør: AAT BIOQUEST
67004
AATB67004EA 1600 NOK
AATB67004
ROX Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Mikroplater PCR-plater
The ROX dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
NB: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3More Product Information