7 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Leverandør: AAT BIOQUEST
67020
AATB67020EA 15925 NOK
AATB67020
7 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Mikroplater PCR-plater
7 dye qPCR calibration plate contains seven spectral calibration plates with seven separate dye standards (TAMRA, SYBR, FAM, JOE, NED, ROX, and VIC).
  • The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
  • This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
NB: Avoid freeze-thawing plates. It is notMore Product Information