Laminar flow cabinets, CrossFlow CLF series

Leverandør: CleanAir by Baker
1560401 1560402 1560403 1560404 1560405 1560406
CLEA1560401EA 0 NOK
CLEA1560401 CLEA1560402 CLEA1560403 CLEA1560404 CLEA1560405 CLEA1560406
Laminar flow cabinets, CrossFlow CLF series
Avtrekksskap og kabinetter Laminert flowbenk
The CrossFlow series is a high quality crossflow cabinet range for handling non-hazardous materials that require a clean working environment. The ergonomically designed HEPA filtered horizontal laminar airflow (crossflow) creates an ISO Class 5 (14644-1) / GMP Class A (Annex 1) work area with low noise level and prevents ambient air from entering the work area, ensuring product protection.

  • Microprocessor flow control
  • User-friendly control panel for intuitively operating
  • HEPA filter H14 (efficiency of 99,995% MPPS) and pre-filter (ISO 16890)
  • Energy saving
More Product Information
Bestill nå

SPESIFIKASJONER

Modell CLF 475 CLF 575 CLF 675
Environmentally Preferable
Lys >1000 lux >1000 lux >1000 lux
Støynivå <58 dB(A) <58 dB(A) <54 dB(A)
Effekt 151 kW 198 kW 245 kW
Filtre HEPA filter H14 accordance with EN 1822 - 1 : 1998 with an efficiency higher than 99,999% on 0,3µ particles, DOP tested
Coarsefilter Pre filter
HEPA filter H14 accordance with EN 1822 - 1 : 1998 with an efficiency higher than 99,999% on 0,3µ particles, DOP tested
Coarsefilter Pre filter
HEPA filter H14 accordance with EN 1822 - 1 : 1998 with an efficiency higher than 99,999% on 0,3µ particles, DOP tested
Coarsefilter Pre filter
Strømforsyning 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz
Utvendig B×D×H 1270×893×1153 mm 1574×893×1153 mm 1880×893×1153 mm
Innvendig B×D×H 1249×570×736 mm 1554×570×736 mm 1859×570×736 mm
Vekt 110 kg 150 kg 190 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR