Vertical laminar flow cabinets, DLF series

Leverandør: CleanAir by Baker
NP0024360 NP0037460 NP0024460 NP0024560 NP0024660 NP0037360
135-0078EA 0 NOK
135-0078 CLEANP0037460 CLEANP0024460 CLEANP0024560 CLEANP0024660 CLEANP0037360
Vertical laminar flow cabinets, DLF series
Avtrekksskap og kabinetter Laminert flowbenk
CleanAir by Baker DLF series is a high quality vertical laminar flow workbench range for handling non-hazardous materials that require a clean working environment. DLF PCR Series is designed to perform PCR. These cabinets ambient air entering the work area, ensures product protection.

  • Microprocessor flow control
  • Smooth interior for efficient and easy cleaning
  • Control control system with alarms and parameters for work mode, UV light, gas valve
  • Epoxy coated steel interior and exterior (1,5 to 2 mm)
  • Stainless Steel worktop
  • Sliding window (safety glass)
More Product Information
Bestill nå

SPESIFIKASJONER

Modell DLF PCR 360 DLF 460 DLF PCR 460 DLF PCR 560 DLF PCR 660 DLF 360
Environmentally Preferable
Lufthastighet 0,35 m/s 0,35 m/s 0,35 m/s 0,35 m/s 0,35 m/s 0,35 m/s
Lys >800 lux >800 lux >800 lux >800 lux >800 lux >800 lux
Støynivå <58 dB(A) <58 dB(A) <58 dB(A) <58 dB(A) <58 dB(A) <58 dB(A)
Filtre Downflow HEPA filter (EN 1822), H14, efficiency efficiency 99,995% MPPS
Pre-filter efficient (EN 779): G3, efficiency 90% Am
Downflow HEPA filter (EN 1822), H14, efficiency efficiency 99,995% MPPS
Pre-filter efficient (EN 779): G3, efficiency 90% Am
Downflow HEPA filter (EN 1822), H14, efficiency efficiency 99,995% MPPS
Pre-filter efficient (EN 779): G3, efficiency 90% Am
Downflow HEPA filter (EN 1822), H14, efficiency efficiency 99,995% MPPS
Pre-filter efficient (EN 779): G3, efficiency 90% Am
Downflow HEPA filter (EN 1822), H14, efficiency efficiency 99,995% MPPS
Pre-filter efficient (EN 779): G3, efficiency 90% Am
Downflow HEPA filter (EN 1822), H14, efficiency efficiency 99,995% MPPS
Pre-filter efficient (EN 779): G3, efficiency 90% Am
Strømforsyning 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz 230 V, 50 Hz
Utvendig B×D×H 985×740×1340 mm 1290×740×1360 mm 1290×740×1360 mm 1595×740×1360 mm 1900×740×1360 mm 985×740×1340 mm
Innvendig B×D×H 885×600×685
mm
1190×600×685 mm 1190×600×685 mm 1495×600×685 mm 1800×600×685 mm 885×600×685
mm
Vekt 140 kg 170 kg 170 kg 200 kg 230 kg 140 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR