Media in bottles, wort agar
Leverandør: DOEHLER GROUP
840360782
DOEH840360782EA 2200 NOK
DOEH840360782
Media in bottles, wort agar
Mikrobiologiske medier Media, preparerte for mikrobiologi
Agar for isolation yeast and moulds.
  • For membrane filtration, pour plate and streaking method
  • High selectivity